missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Aminocyclohexanol hydrochloride, 99+%, Thermo Scientific Chemicals

442.20 € - 1081.38 €

Chemical Identifiers

CAS 6936-47-6
MDL Number MFCD00143981
InChI Key LKKCSUHCVGCGFA-RIHPBJNCSA-N
Synonym 1r,2s-2-aminocyclohexanol hydrochloride, cis-2-aminocyclohexanol hydrochloride, 1r,2s-2-aminocyclohexan-1-ol hydrochloride, 1r,2s-rel-2-aminocyclohexanol hydrochloride, cis 1r,2s-2-amino-cyclohexanol hydrochloride, 1r,2s-2-aminocyclohexanol hcl, cyclohexanol, 2-amino-, hydrochloride, cis, cis-2-aminocyclohexanolhydrochloride, cis-2-aminocyclohexanol hcl, cis-2-amino-cyclohexanol hydrochloride
PubChem CID 12228413
IUPAC Name (1R,2S)-2-aminocyclohexan-1-ol;hydrochloride
SMILES C1CCC(C(C1)N)O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10389000
View Documents
Thermo Scientific Acros
265850010
1 g
442.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
10722964
View Documents
Thermo Scientific Acros
265850050
5 g
1081.38 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6936-47-6
LKKCSUHCVGCGFA-RIHPBJNCSA-N
12228413
C1CCC(C(C1)N)O.Cl
MFCD00143981
1r,2s-2-aminocyclohexanol hydrochloride, cis-2-aminocyclohexanol hydrochloride, 1r,2s-2-aminocyclohexan-1-ol hydrochloride, 1r,2s-rel-2-aminocyclohexanol hydrochloride, cis 1r,2s-2-amino-cyclohexanol hydrochloride, 1r,2s-2-aminocyclohexanol hcl, cyclohexanol, 2-amino-, hydrochloride, cis, cis-2-aminocyclohexanolhydrochloride, cis-2-aminocyclohexanol hcl, cis-2-amino-cyclohexanol hydrochloride
(1R,2S)-2-aminocyclohexan-1-ol;hydrochloride

Specifications

6936-47-6
White to Beige
99+%
C6H13NO·HCl
MFCD00143981
Solubility in water: soluble. Other solubilities: soluble in methanol,hot ethanol
C1CCC(C(C1)N)O.Cl
151.64
151.64
Crystalline Powder
186.0°C to 190.0°C
Authentic
Glass bottle
H2NC6H10OH·HCl
1r,2s-2-aminocyclohexanol hydrochloride, cis-2-aminocyclohexanol hydrochloride, 1r,2s-2-aminocyclohexan-1-ol hydrochloride, 1r,2s-rel-2-aminocyclohexanol hydrochloride, cis 1r,2s-2-amino-cyclohexanol hydrochloride, 1r,2s-2-aminocyclohexanol hcl, cyclohexanol, 2-amino-, hydrochloride, cis, cis-2-aminocyclohexanolhydrochloride, cis-2-aminocyclohexanol hcl, cis-2-amino-cyclohexanol hydrochloride
LKKCSUHCVGCGFA-RIHPBJNCSA-N
(1R,2S)-2-aminocyclohexan-1-ol;hydrochloride
12228413
99+%
cis-2-Aminocyclohexanol hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.