Learn More
cis-3-Methylcyclohexanol, 95%, Thermo Scientific Chemicals
116.58 € - 439.00 €
Chemical Identifiers
CAS | 24965-90-0 |
---|---|
Molecular Formula | C7H14O |
Molecular Weight (g/mol) | 114.188 |
MDL Number | MFCD00067155 |
InChI Key | HTSABYAWKQAHBT-RQJHMYQMSA-N |
Synonym | cis-3-methylcyclohexanol, cyclohexanol, 3-methyl-, cis, 1s,3r-3-methylcyclohexan-1-ol, 3-methyl cyclohexanol, cis, 3-methylcyclohexanol, z, cis 3-methylcyclohexanol, cis-3-methyl-cyclohexanol, 1s,3r-3-methyl-cyclohexanol, 1alpha-3alpha-methylcyclohexanol |
PubChem CID | 229503 |
IUPAC Name | (1S,3R)-3-methylcyclohexan-1-ol |
SMILES | CC1CCCC(C1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429796
|
Thermo Scientific Alfa Aesar
B20288.06 |
5 g |
116.58 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439796
|
Thermo Scientific Alfa Aesar
B20288.14 |
25 g | N/A | ||||||
Description
cis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationscis-3-Methylcyclohexanol is a useful cis methyl cyclohexanol for proteomics research and as an intermediate in organic synthesis.
Solubility
Soluble in ethanol, ether. Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
24965-90-0 | |
114.188 | |
HTSABYAWKQAHBT-RQJHMYQMSA-N | |
229503 | |
CC1CCCC(C1)O |
C7H14O | |
MFCD00067155 | |
cis-3-methylcyclohexanol, cyclohexanol, 3-methyl-, cis, 1s,3r-3-methylcyclohexan-1-ol, 3-methyl cyclohexanol, cis, 3-methylcyclohexanol, z, cis 3-methylcyclohexanol, cis-3-methyl-cyclohexanol, 1s,3r-3-methyl-cyclohexanol, 1alpha-3alpha-methylcyclohexanol | |
(1S,3R)-3-methylcyclohexan-1-ol |
Specifications
24965-90-0 | |
C7H14O | |
cis-3-methylcyclohexanol, cyclohexanol, 3-methyl-, cis, 1s,3r-3-methylcyclohexan-1-ol, 3-methyl cyclohexanol, cis, 3-methylcyclohexanol, z, cis 3-methylcyclohexanol, cis-3-methyl-cyclohexanol, 1s,3r-3-methyl-cyclohexanol, 1alpha-3alpha-methylcyclohexanol | |
HTSABYAWKQAHBT-RQJHMYQMSA-N | |
(1S,3R)-3-methylcyclohexan-1-ol | |
229503 | |
95% |
73°C (163°F) | |
MFCD00067155 | |
Soluble in ethanol,ether. Slightly soluble in water. | |
CC1CCCC(C1)O | |
114.188 | |
114.19 | |
cis-3-Methylcyclohexanol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.