missing translation for 'onlineSavingsMsg'
Learn More
cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals Product Code.: 15429328

cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 15429328
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15429328

Brand: Thermo Scientific Alfa Aesar H62871.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

cis-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexanecarboxaldehyde is used as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181308-56-5
Molecular Formula C12H21NO3
Molecular Weight (g/mol) 227.304
MDL Number MFCD03844599
InChI Key GPDBIGSFXXKWQR-UHFFFAOYSA-N
Synonym tert-butyl trans-4-formylcyclohexylcarbamate, tert-butyl cis-4-formylcyclohexylcarbamate, trans-boc-4-aminocyclohexanecarbaldehyde, tert-butyl n-4-formylcyclohexyl carbamate, trans-4-formyl-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-formylcyclohexyl carbamate, tert-butyl 4-formylcyclohexyl carbamate, trans-tert-butyl 4-formylcyclohexyl carbamate, tert-butyl 1r,4r-4-formylcyclohexylcarbamate, cis-4-boc-amino cyclohexanecarboxaldehydeShow MoreShow Less
PubChem CID 11020615
IUPAC Name tert-butyl N-(4-formylcyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C=O

Specifications

Quantity 5 g
Formula Weight 227.3
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexanecarboxaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP06J040 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberH62871.03,H62871.06
Lot NumberQ25C014 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62871.03,H62871.06
Lot NumberN12F021 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62871.03,H62871.06
Lot NumberC29Z007 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62871.03,H62871.06
Lot NumberT01C003 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH62871.03,H62871.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
cis-4-(Boc-amino)cyclohexanecarboxaldehyde, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.