missing translation for 'onlineSavingsMsg'
Learn More

cis-4-(Boc-amino)cyclohexanol, 97%, Thermo Scientific Chemicals

Product Code. 15448788
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents

Solubility
Slightly soluble in water

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167081-25-6
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
MDL Number MFCD03844613,MFCD03844614,MFCD06658349
InChI Key DQARDWKWPIRJEH-UHFFFAOYSA-N
Synonym trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate
PubChem CID 1514287
IUPAC Name tert-butyl N-(4-hydroxycyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(O)CC1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water
Formula Weight 215.28
Percent Purity 97%
Chemical Name or Material cis-4-(Boc-amino)cyclohexanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'