Learn More
cis-4-(Boc-amino)cyclohexanol, 97%, Thermo Scientific Chemicals
55.60 € - 686.00 €
Chemical Identifiers
CAS | 167081-25-6 |
---|---|
Molecular Formula | C11H21NO3 |
Molecular Weight (g/mol) | 215.29 |
MDL Number | MFCD03844613,MFCD03844614,MFCD06658349 |
InChI Key | DQARDWKWPIRJEH-UHFFFAOYSA-N |
Synonym | trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate |
PubChem CID | 1514287 |
IUPAC Name | tert-butyl N-(4-hydroxycyclohexyl)carbamate |
SMILES | CC(C)(C)OC(=O)NC1CCC(O)CC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448788
|
Thermo Scientific Alfa Aesar
H62238.MD |
250 mg | |||||||
15428788
|
Thermo Scientific Alfa Aesar
H62238.03 |
1 g |
176.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15438788
|
Thermo Scientific Alfa Aesar
H62238.06 |
5 g |
686.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
cis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationscis-4-(Boc-amino)cyclohexanol is used as a reagent to synthesize pyrazolo[3,4-d]pyrimidines, compounds that potentially act as anti-inflammatory and antitumor agents
Solubility
Slightly soluble in water
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
167081-25-6 | |
215.29 | |
DQARDWKWPIRJEH-UHFFFAOYSA-N | |
1514287 | |
CC(C)(C)OC(=O)NC1CCC(O)CC1 |
C11H21NO3 | |
MFCD03844613,MFCD03844614,MFCD06658349 | |
trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate | |
tert-butyl N-(4-hydroxycyclohexyl)carbamate |
Specifications
167081-25-6 | |
MFCD03844613,MFCD03844614,MFCD06658349 | |
Slightly soluble in water | |
CC(C)(C)OC(=O)NC1CCC(O)CC1 | |
215.29 | |
215.28 | |
cis-4-(Boc-amino)cyclohexanol |
C11H21NO3 | |
trans-4-boc-aminocyclohexanol, tert-butyl 4-hydroxycyclohexyl carbamate, boc-trans-4-aminocyclohexanol, 4-boc-amino cyclohexanol, tert-butyl trans-4-hydroxycyclohexyl carbamate, tert-butyl n-4-hydroxycyclohexyl carbamate, tert-butyl cis-4-hydroxycyclohexylcarbamate, trans-n-boc-4-amino-cyclohexanol, 4-n-boc-amino-cyclohexanol, tert-butyl cis-4-hydroxycyclohexyl carbamate | |
DQARDWKWPIRJEH-UHFFFAOYSA-N | |
tert-butyl N-(4-hydroxycyclohexyl)carbamate | |
1514287 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only