missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cis-5,8,11,14,17-Eicosapentaenoic acid

Product Code. 15497269
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15497269

Brand: Thermo Scientific Chemicals J66060.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10417-94-4 | C20H30O2, C20H30O2 | 302.46 g/mol

Important polyunsaturated fatty acid of the marine food chain that serves as a precursor for the prostaglandin-3 and thromboxane-3 families. It differs from arachidonic acid (the eicosatetraenoic acid that is a precursor for the prostaglandin and thromboxane-2 families) by the extra double bond between the third and fourth carbons from the methyl end of the molecule. Antilipemic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10417-94-4
Molecular Formula C20H30O2
Synonym EPA; Timnodonic acid

Specifications

Quantity 100 mg
Description Colorless
Content And Storage -30°C to -10°C
Formula Weight 302.45
Flash Point 93°C (199°F)
Density 0.943
Sensitivity Air and Light Sensitive
Melting Point -54°
Solubility Soluble in methanol
Product Type cis-5,8,11,14,17-Eicosapentaenoic acid
Form Oil
Beilstein 1714433
InChI Key JAZBEHYOTPTENJ-JLNKQSITSA-N
SMILES CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(O)=O
IUPAC Name (5Z,8Z,11Z,14Z,17Z)-icosa-5,8,11,14,17-pentaenoic acid
Molecular Weight (g/mol) 302.46
Show More Show Less
compliance-icons
Product Identifier
  • cis-5,8,11,14,17-Eicosapentaenoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Eicosapentaenoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.