missing translation for 'onlineSavingsMsg'
Learn More

cis,cis-1,3,5-Cyclohexanetriol dihydrate, 98%, Thermo Scientific Chemicals

Product Code. 15590594
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60662-54-6
Molecular Formula C6H12O3·2H2O
Molecular Weight (g/mol) 168.19
InChI Key MAPLMYMZLRIJSP-UHFFFAOYSA-N
Synonym cis,cis-1,3,5-cyclohexanetriol dihydrate, cis,cis-1,3,5-trihydroxycyclohexane, cis-phloroglucitol, 1s,3s,5s-cyclohexane-1,3,5-triol dihydrate, ciscis-135-cyclohexanetrioldihydrate, cyclohexane-1,3,5-triol dihydrate, cyclohexane-1,3,5-triol-water 1/2, cis cis-1 3 5-cyclohexanetriol dihydrate, 1r,3s,5s-cyclohexane-1,3,5-triol dihydrate
PubChem CID 16211857
IUPAC Name cyclohexane-1,3,5-triol;dihydrate
SMILES C1C(CC(CC1O)O)O.O.O

Specifications

Melting Point 108.0°C to 110.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble in hot water.
Formula Weight 168.19
Percent Purity 98%
Chemical Name or Material cis,cis-1,3,5-Cyclohexanetriol dihydrate
Product Identifier
  • cis,cis-1,3,5-Cyclohexanetriol dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'