missing translation for 'onlineSavingsMsg'
Learn More

CIT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen CIT
Concentration 0.37 mg/mL
Applications Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16899514
View Documents
Proteintech
20033-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16889514
View Documents
Proteintech
20033-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The antibody recognizes all the isoforms of CIT.

This gene encodes a serine/threonine-protein kinase that functions in cell division. Together with the kinesin KIF14, this protein localizes to the central spindle and midbody, and functions to promote efficient cytokinesis. This protein is involved in central nervous system development. Polymorphisms in this gene are associated with bipolar disorder and risk for schizophrenia. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

CIT
Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
O14578, P49025
11113, 12704, 83620
Peptide
Primary
-20°C
CIT
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CIT, Citron Rho interacting kinase, CRIK, KIAA0949, STK21
Cit
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications