Learn More
Citric Acid, Anhydrous, 99%, Pure, Thermo Scientific Chemicals
61.41 € - 553.76 €
Chemical Identifiers
CAS | 77-92-9 |
---|---|
Molecular Formula | C6H8O7 |
Molecular Weight (g/mol) | 192.12 |
MDL Number | MFCD00011669 |
InChI Key | KRKNYBCHXYNGOX-UHFFFAOYSA-N |
Synonym | citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy |
PubChem CID | 311 |
ChEBI | CHEBI:30769 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid |
SMILES | OC(=O)CC(O)(CC(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10020760
|
Thermo Scientific Acros
110450010 |
1 kg | Plastic Bottle |
61.41 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10338903
|
Thermo Scientific Acros
110450050 |
5 kg | Plastic Bottle |
156.78 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10094003
|
Thermo Scientific Acros
110450250 |
25 kg | Fibre drum |
553.76 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
77-92-9 | |
192.12 | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
311 | |
2-hydroxypropane-1,2,3-tricarboxylic acid |
C6H8O7 | |
MFCD00011669 | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
CHEBI:30769 | |
OC(=O)CC(O)(CC(O)=O)C(O)=O |
Specifications
77-92-9 | |
White | |
C6H8O7 | |
MFCD00011669 | |
15, 2325 | |
Solubility in water: 750g/L (20°C). Other solubilities: soluble in alcohol and ether, badly soluble in diethyl ether, insoluble in benzene and chloroform | |
OC(=O)CC(O)(CC(O)=O)C(O)=O | |
192.12 | |
CHEBI:30769 | |
99% | |
100°C | |
Crystalline |
153.0°C | |
99% | |
HO2CCH2C(OH)(CO2H)CH2CO2H | |
03, 556 | |
citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy | |
KRKNYBCHXYNGOX-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid | |
311 | |
192.13 | |
Pure | |
Authentic | |
Citric acid, Anhydrous |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Citric acid anhydrous
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.