missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Anhydrous (Crystalline), Fisher BioReagents

Product Code. 10193333
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Anhydrous citric acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

pH 1.7
CAS 77-92-9
Melting Point 153°C
Color White
Assay Percent Range ≥99.5 %
Molecular Formula C6H8O7
MDL Number MFCD00011669
Merck Index 15, 2325
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
PubChem CID 311
ChEBI CHEBI:30769
Formula Weight 192.13
Percent Purity ≥99.5%
Quantity 500 g
Ignition Residue 0.02% max.
Packaging Poly Bottle
Physical Form Solid
Chemical Name or Material Citric Acid Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Citric Acid Anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.