missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid, Anhydrous, Grit, Honeywell™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
76.95 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 77-92-9 |
Molecular Formula | C6H8O7 |
Linear Formula | HOC(COOH)(CH2COOH)2 |
MDL Number | MFCD00011669 |
UN Number | NONH for all modes of transport |
Beilstein | 782061 |
Synonym | citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy |
Solubility Information | Very slightly soluble in diethyl ether, freely soluble in alcohol, Soluble in water 383g/L at 25°C |
InChI Key | KRKNYBCHXYNGOX-UHFFFAOYSA-N |
SMILES | OC(=O)CC(O)(CC(O)=O)C(O)=O |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'