Learn More
Citric acid monohydrate, 99.5+%, Thermo Scientific Chemicals
26.83 € - 102.51 €
Chemical Identifiers
CAS | 5949-29-1 |
---|---|
Molecular Formula | C6H10O8 |
Molecular Weight (g/mol) | 210.138 |
MDL Number | MFCD00149972 |
InChI Key | YASYEJJMZJALEJ-UHFFFAOYSA-N |
Synonym | citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp |
PubChem CID | 22230 |
ChEBI | CHEBI:31404 |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348107
|
Thermo Scientific Alfa Aesar
022869.22 |
100 g |
26.83 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328107
|
Thermo Scientific Alfa Aesar
022869.36 |
500 g |
54.77 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11338107
|
Thermo Scientific Alfa Aesar
022869.A1 |
1 kg |
102.51 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Citric acid monohydrate is used in the preparation of citrate buffer in platelets for intravital microscopy. It acts as a pH-control agent in foods, beverages and pharmaceuticals applications. It acts as an iron chelator. In animals, it improves the utilization of nutritional calcium. It is a useful buffer component for antigen and epitope unmasking. It is also used as an acidifier, flavoring agent and chelating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5949-29-1 | |
210.138 | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
22230 | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate |
C6H10O8 | |
MFCD00149972 | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
CHEBI:31404 | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O |
Specifications
5949-29-1 | |
C6H10O8 | |
14,2326 | |
Soluble in water,ethanol and ether. | |
C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O | |
210.138 | |
CHEBI:31404 | |
≥99.5% | |
Odorless | |
Hygroscopic |
White | |
MFCD00149972 | |
citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate usp | |
YASYEJJMZJALEJ-UHFFFAOYSA-N | |
2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate | |
22230 | |
210.14 (192.13 Anhydrous) | |
1.542 g/mL | |
Granular Powder | |
Citric acid monohydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280i-P305+P351+P338
H319-H500
missing translation for 'einecsNumber' : 201-069-1
missing translation for 'rtecsNumber' : GE7810000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.