missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid Monohydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™

Product Code. 10345410
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar

C6H8O7.H2O, CAS Number-5949-29-1, 5kg, White, 210.15g/mol, 135 deg.C, 149972, Plastic keg, 1.8, Solid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.14
MDL Number 149972
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
SMILES O.OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 1.8
Melting Point 135°C
Color White
Formula Weight 210.15g/mol
Quantity 500 g
Packaging Plastic powder jar
Physical Form Solid
compliance-icons
Product Identifier
  • Citric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash A§ thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.