missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Monohydrate, HPLC for Electrochemical Detection, Certified, Fisher Chemical
Click to view available options
500g, Plastic powder jar
Description
500g, Plastic powder jar
Specifications
Specifications
pH | 1.8 |
CAS | 5949-29-1 |
Melting Point | 135°C |
Color | White |
Molecular Formula | C6H10O8 |
MDL Number | 149972 |
InChI Key | YASYEJJMZJALEJ-UHFFFAOYSA-N |
SMILES | O.OC(=O)CC(O)(CC(O)=O)C(O)=O |
IUPAC Name | 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate |
Molecular Weight (g/mol) | 210.14 |
Show More |
Safety and Handling
Product Identifier
- Citric acid monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'