missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, trisodium salt dihydrate, 99%, pure, Thermo Scientific Chemicals

Product Code. 13438488
Click to view available options
Quantity:
5 g
500 g
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Glass Bottle
Plastic Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1545801
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.1
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Fibre drum
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
Quantity 25 kg
Merck Index 15, 8737
Solubility Information Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol
Formula Weight 294.09
Percent Purity 0.99
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Citric acid, trisodium salt dihydrate
Show More Show Less
Product Identifier
  • Citric acid, trisodium salt dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'