missing translation for 'onlineSavingsMsg'
Learn More

Cl-4AS-1, Tocris Bioscience™

Product Code. 18083792
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Steroidal androgen receptor agonist

Potent steroidal androgen receptor agonist (IC50 = 12 nM). Mimics the action of 5α-dihydrotestosterone (DHT). Transactivates the mouse mammary tumor virus (MMTV) promoter; represses MMP1 promoter activity. Inhibits 5α-reductase type I and II (IC50 values are 6 and 10 nM respectively).

Chemical Identifiers

CAS 188589-66-4
Molecular Formula C26H33ClN2O2
Molecular Weight (g/mol) 441.012
InChI Key CTVXDPDUOKQBKZ-BLIBDJTASA-N
Synonym cl-4as-1, 4a?,4b?,6a?,7?,9a?,9b?,11a?-n-2-chlorophenyl-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-1,4,6a-trimethyl-2-oxo-1h-indeno 5,4-f quinoline-7-carboxamide
PubChem CID 90488881
IUPAC Name (1S,3aS,3bS,9aR,9bS,11aR)-N-(2-chlorophenyl)-6,9a,11a-trimethyl-7-oxo-2,3,3a,3b,4,5,5a,9b,10,11-decahydro-1H-indeno[5,4-f]quinoline-1-carboxamide
SMILES CC12CCC3C(C1CCC2C(=O)NC4=CC=CC=C4Cl)CCC5C3(C=CC(=O)N5C)C

Specifications

Quantity 10 mg
Formula Weight 441.01
Percent Purity >99%
Chemical Name or Material Cl-4AS-1
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'