Learn More
Heidolph™ Clamp for Erlenmeyer Flasks
Clamps for use with various perforated platforms
20.50 € - 71.75 €
Product Code | Brand | Size | Holds | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Holds | For Use With (Equipment) | Price | Quantity & Availability | |||
10242501
|
Heidolph™
549-51000-00 |
1 | 25 mL Erlenmeyer flasks | Perforated platforms 100, 1000 and 2000 |
20.50 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
|||
10739171
|
Heidolph™
549-52000-00 |
2 | 50 mL Erlenmeyer flasks | Perforated platforms 100, 1000 and 2000 |
30.75 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
10749171
|
Heidolph™
549-53000-00 |
3 | 100 mL Erlenmeyer flasks | Perforated platforms 100, 1000 and 2000 |
30.75 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
10151361
|
Heidolph™
549-54000-00 |
4 | 250 mL Erlenmeyer flasks | Perforated platforms 100, 1000 and 2000 |
41.00 €
Each |
Estimated Shipment: 08-01-2025
Log in to see stock. |
|||
10789551
|
Heidolph™
549-55000-00 |
5 | 500 mL Erlenmeyer flasks | Perforated platforms 100, 1000 and 2000 |
41.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||
10099340
|
Heidolph™
549-56000-00 |
6 | 1000 mL Erlenmeyer flasks | Perforated platforms 100, 1000 and 2000 |
51.25 €
Each |
Estimated Shipment: 31-12-2024
Log in to see stock. |
|||
11711659
|
Heidolph™
549-63000-00 |
7 | 2000 mL Erlenmeyer flasks | Perforated platforms 1000 and 2000 |
71.75 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
Description
- Maximum number of clamps per platform type:
- 549-51000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-52000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-55000-00
- For platform 100 - max. 3
- For platform 1000 - max. 4
- For platform 2000 - max. 9
- 549-56000-00
- For platform 100 - max. 2
- For platform 1000 - max. 4
- For platform 2000 - max. 5
- 549-63000-00
- For platform 1000 - max. 2
- For platform 2000 - max. 3
- 549-51000-00
Specifications
Clamp |