Learn More
DWK Life Sciences Clamp for Spherical Joints, with tightening screw, made of stainless steel
Ball joint clamp with additional locking option.
Brand:  DWK Life Sciences 286334605
 View more versions of this product
Product Code. 15844678
  • 713.00 € / Pack of 25
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The ball joint clamp with locking screw enables a very secure connection by additional locking with the locking screw.
  • The locking screw enables a very secure connection by means of an additional locking device.
  • Excellent dimensional stability compared to conventional wire clamps
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25
Securing of Spherical Ground Joints
S 40 and S 41
Stainless Steel
Securing of spherical ground joints