Learn More
Kartell™ Clamps for Glass Joints
Plastic clamps for safe connection of standard-finish glass conical joints
Brand:  Kartell™ 3101900
 View more versions of this product
Product Code. 15875577
  • 31.55 € / Pack of 10
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Corrosion proof
  • Do not scratch glass
  • Available in various colours according to dimension
  • Autoclavable
Specifications
Specifications
10/Pk.
POM
10/19
Black