missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Class 3.1 Independent Temperature Safety Device

Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)

605.00 €

Specifications

Description Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
Type Temperature Calibration Certificate
For Use With (Application) Temperature Regulation
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
16454349
View Documents
BINDER™
8012-1643
BINDER Chambers FD 56, FD 115, FD 260, BINDER Chambers FD 720
605.00 €
Each
Estimated Shipment: 10-03-2025
to see stock.
Add to basket
 
16464349
View Documents
BINDER™
8012-1644
BINDER Chambers FED 56, FED 115, FED 260, FED 720
605.00 €
Each
Estimated Shipment: 15-04-2025
to see stock.
Add to basket
 
Description

Description

  • Class 3.1
  • With visual alarm
  • Safety devices also available in Class 2 and 3.3
Specifications

Specifications

Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
Temperature Calibration Certificate
Temperature Regulation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.