Learn More
Manutan Class 3 flammable liquid IATA/ICAO labels, 100mm x 100mm
Brand:  Manutan A352629
Encompass_Preferred
Product Code. 12818948
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
A range of unique labels with varying class numbers. Universally agreed airline standards to identify and safely transport dangerous goods by air. Used and recognised by all major carriers and forwarders. Printed on self-adhesive vinyl tape, with clear polypropylene over-laminate.
  • Flammable liquid
  • Self-adhesive vinyl
  • Dimensions (L x H) : 100mm x 100mm
  • Class 3