Learn More
Hirschmann™ Class A Brown Graduated USP Measuring Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs.
Brand:  Hirschmann™ 134017527
 View more versions of this product
Product Code. 11751526
  • 94.75 € / Pack of 6
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With dated batch identification
  • Manufactured from AR glass™ soda-lime glass with a smooth, non-porous surface
  • With volume mark
  • Brown graduation
Specifications
Specifications
A
Volumetric accuracy in pipetting
Brown
50 mL
AR-GLAS
Yes
0.05 mL
540 ±10 mm