Learn More
Hirschmann™ Class A Kohlrausch Measuring Flasks
For sugar examination pursuant to Kohlrausch.
Brand:  Hirschmann™ 2970180
Product Code. 11720854
  • 59.45 € / Pack of 2
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class A
  • Manufactured from DURAN™ borosilicate glass for resistance to chemicals and thermal shock
  • Certified conformity, with dated batch identification
  • Calibrated for IN
  • Blue graduations
Specifications
Specifications
Calibrated for IN
Borosilicate Glass
Blue
Measuring flask
Yes
Flat
100 mL
Sugar examination
170 ± 5 mm
0.1 mL
A