Learn More
Hirschmann™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket are manufactured from durable DURAN™ glass tubing.
22.05 € - 81.20 €
Specifications
Base Type Hexagonal
Certifications/Compliance DIN EN ISO 4788
Class A
Type Graduated Measuring Cylinder
Scale Color Blue
View More Specs
Products 7
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11756692
View accessory products View Documents
Hirschmann™
2270160
10 mL
22.05 €
Pack of 2
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
11766692
View accessory products View Documents
Hirschmann™
2270170
25 mL
22.65 €
Pack of 2
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
11776692
View accessory products View Documents
Hirschmann™
2270175
50 mL
26.90 €
Pack of 2
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
11756702
View accessory products View Documents
Hirschmann™
2270180
100 mL
29.05 €
Pack of 2
In Stock
to see stock.
Add to basket
11766702
View accessory products View Documents
Hirschmann™
2270186
250 mL
50.95 €
Pack of 2
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
11776702
View accessory products View Documents
Hirschmann™
2270190
500 mL
81.20 €
Pack of 2
Estimated Shipment: 10-01-2025
to see stock.
Add to basket
10557961
View Documents
Hirschmann™
2270193
1000 mL
63.90 €
Each
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
Description
Description
  • With spout and plastic socket, main point ring graduation
  • Blue graduation
  • Conformity certified, with dated batch information
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Hexagonal
A
Blue
Glass
DIN EN ISO 4788
Graduated Measuring Cylinder
Round