Learn More
Hirschmann™ Class A Measuring Cylinders, Amber Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Amber Graduation are manufactured from durable DURAN™ glass tubing.
28.00 € - 174.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10542404
|
Hirschmann™
2210154 |
5 mL |
28.00 €
Pack of 2 |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
13117939
|
Hirschmann™
2210160 |
10 mL |
33.15 €
Pack of 2 |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
10117411
|
Hirschmann™
2210170 |
25 mL |
35.20 €
Pack of 2 |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
12751528
|
Hirschmann™
2210175 |
50 mL |
40.45 €
Pack of 2 |
In Stock
Log in to see stock. |
|||||
10505921
|
Hirschmann™
2210180 |
100 mL |
40.20 €
Pack of 2 |
In Stock
Log in to see stock. |
|||||
10479662
|
Hirschmann™
2210186 |
250 mL |
65.60 €
Pack of 2 |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
11834382
|
Hirschmann™
2210190 |
500 mL |
133.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||||
11874412
|
Hirschmann™
2210193 |
1000 mL |
98.00 €
Each |
In Stock
Log in to see stock. |
|||||
10262431
|
Hirschmann™
2210194 |
2000 mL |
174.00 €
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
- With spout, main point ring amber stain graduation
- Conformity certified, with dated batch identification
- Acc. to DIN EN ISO 4788
- Corrosion-free and resistant to thermal shock
Specifications
DIN EN ISO 4788 | |
A | |
Glass |