Learn More
Hirschmann™ AR™ Glass Volumetric Pipettes
Provides an extremely high degree of volumetric accuracy for your pipetting needs. Hirschmann Instruments™ AR™ Glass Volumetric Pipettes have a tolerance better than Class B.
Brand:  Hirschmann™ 1300160
 View more versions of this product
Product Code. 11787971
  • 50.15 € / Pack of 6
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • One mark
  • Amber stain graduation
  • Acc. to DIN EN ISO 648
Specifications
Specifications
B
Yes
6/Pk.
10 mL
Volumetric Pipet
Glass
Brown
0.03 mL
450 mm