Learn More
Hecht Karl™ Class B DURAN Borosilicate Glass Tall Form Measuring Cylinders
Brand:  Hecht Karl™ 42170072
 View more versions of this product
Product Code. 10003921
  • 91.00 € / Each
Explore more special offers
Specifications
±15 mL
Blue
B
Blue Graduations
Graduated Measuring Cylinder
Borosilicate Glass
DIN EN ISO 4788
2000 mL
±15mL
±15mL