missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zwiebel™ Class E2 Weight with COFRAC Calibration Certificate
Calibrate weights of lower accuracy classes. Zwiebel™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
241.00 €
Specifications
For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |
---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Tolerance | Price | Quantity & Availability | ||||
11700643
|
Zwiebel™
PBI2302S |
100 mg | ±0.016 mg |
241.00 €
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
11770633
|
Zwiebel™
PBI2302N |
10 mg | ±0.008 mg |
241.00 €
Each |
Estimated Shipment: 31-01-2025
Log in to see stock. |
||||
11710643
|
Zwiebel™
PBI2302T |
200 mg | ±0.02 mg |
241.00 €
Each |
Estimated Shipment: 11-02-2025
Log in to see stock. |
||||
11740633
|
Zwiebel™
PBI2302K |
1 mg | ±0.006 mg |
241.00 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
11720643
|
Zwiebel™
PBI2302W |
500 mg | ±0.025 mg |
241.00 €
Each |
Estimated Shipment: 13-02-2025
Log in to see stock. |
||||
11780633
|
Zwiebel™
PBI2302P |
20 mg | ±0.01 mg |
241.00 €
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
||||
11790633
|
Zwiebel™
PBI2302R |
50 mg | ±0.012 mg |
241.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||||
Description
- Class E2 calibration weight
- In compliance with OIML R111
- Delivered with a COFRAC calibration certificate guaranteeing traceability
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
For use with Weighing Instruments of Special Accuracy Class I |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title