missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Box Weights

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.

Brand:  Zwiebel™ PBI2300C

 View more versions of this product

Product Code. 11700593

  • 19.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class M1 weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications

    Specifications

    ±0.2 mg
    For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers