Learn More
Zwiebel™ Class M1 Weight Boxes with Certificate
Brand:  Zwiebel™ PBI56810
 View more versions of this product
Product Code. 11890882
  • 158.00 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
500 g
±25 mg
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII