Learn More
Abeba™ Classic 31010 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
89.10 €
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB SRC
Color Black
Liner Material Microfiber
Material Leather
Sole Features Slip-resistant
View More Specs
Products 9
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15658277
View Documents
Abeba™
31010/38
38
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15668277
View Documents
Abeba™
31010/39
39
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15678277
View Documents
Abeba™
31010/40
40
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15688277
View Documents
Abeba™
31010/41
41
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15698277
View Documents
Abeba™
31010/42
42
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15608287
View Documents
Abeba™
31010/43
43
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15618287
View Documents
Abeba™
31010/44
44
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15628287
View Documents
Abeba™
31010/45
45
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15638287
View Documents
Abeba™
31010/46
46
89.10 €
1 pair
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 SB SRC
  • Grained leather black
  • Steel toe cap
  • Slip resistance PU outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • Buckle fastening
  • Heel patch microfiber
Specifications
Specifications
CE EN ISO 20345:2011 SB SRC
Microfiber
Slip-resistant
31010
Animal fats, oil and fuel
Black
Leather
Polyurethane
Steel