Learn More
Cytiva Classical Powdered Ham's Nutrient Mixture F10 - F12
Brand:  Cytiva SH30010.04
 View more versions of this product
Product Code. 10733374
  • 183.00 € / 50 L
Explore more special offers
Description
Description
Support cell culture growth and biomanufacturing processes with powdered HyClone™ Ham's F10-F12 Nutrient Mixture, manufactured using ISO 9001:2000-certified processes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
cGMP and ISO 9001:2000
Ham's F12, with L-Glutamine
Ham's F12, with L-glutamine
Glutamine
1 x 50 L
2° to 8°C, away from light
Powder
Non-sterile
No Sodium Bicarbonate