Learn More
Brannan™ Classroom Wall Kit - Thermometer and Chart with 3 Point Traceable Calibration Certificate
Single packed classroom wall thermometer kit with laminated chart. Complete with 3 point traceable calibration.
Brand:  Brannan™ 25/702/0 + 49/101/0
Product Code. 17842988
  • 155.00 € / Each
Explore more special offers
Description
Description
  • Colorful 380 mm plastic wall thermometer with chart for recording temperatures
  • Brightly designed, ideal for education
  • Supplied with calibration certificate
  • Laminated chart making in easy to clean for multiple uses
  • Made in UK
Specifications
Specifications
UK National Standards ISO 9001
Wall Thermometer with chart
-20 to +50°C
Wall Thermometer Kit