missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Clay S3 Shoe

If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay.

133.00 € - 154.00 €

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
View More Specs

Products 8
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17582626
View Documents
Emma Safety Footwear
MM98054939
39
154.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17592626
View Documents
Emma Safety Footwear
MM98054940
40
133.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17502636
View Documents
Emma Safety Footwear
MM98054941
41
133.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17512636
View Documents
Emma Safety Footwear
MM98054942
42
139.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17522636
View Documents
Emma Safety Footwear
MM98054945
45
133.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17532636
View Documents
Emma Safety Footwear
MM98054946
46
133.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17542636
View Documents
Emma Safety Footwear
MM98054947
47
133.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
17552636
View Documents
Emma Safety Footwear
MM98054948
48
133.00 €
1 pair
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
Description

Description

  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay. A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel.
  • This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications

Specifications

ISO 20345:2011
Black
Hydro-Tec Antibacterial
Standard Width
Safety Shoe
Laces
Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Nubuck, Oiled Nubuck
Low-cut
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.