missing translation for 'onlineSavingsMsg'
Learn More

Scotch-Brite™ Clean & Shine Double Sided Floor Pads

This 2 in 1 daily cleaning pad for low speed scrubbers gradually increases shine with repeated use

Brand:  Scotch-Brite™ 7100147921

 View more versions of this product

Product Code. 17168760

  • 200.00 € / Pack of 5
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cleans and shines in the same step
  • For use with a low-speed scrubber
  • Save costs and labor by reducing or eliminating the need to burnish
  • Removes black marks 3 times faster than traditional cleaning pads
  • Two-sided pad lasts longer than traditional cleaning pads
  • Effective on most coated and uncoated hard floors including VCT, vinyl, rubber, marble, stone, terrazzo, granite, and concrete with only water or neutral cleaner
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cleaning Pads
305 mm
Low Speed Scrubbers
Brown
Cleaning and Polishing
2 in. 1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.