missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scotch-Brite™ Clean & Shine Double Sided Floor Pads
This 2 in 1 daily cleaning pad for low speed scrubbers gradually increases shine with repeated use
Brand: Scotch-Brite™ 7100148006
This item is not returnable.
View return policy
Description
- Cleans and shines in the same step
- For use with a low-speed scrubber
- Save costs and labor by reducing or eliminating the need to burnish
- Removes black marks 3 times faster than traditional cleaning pads
- Two-sided pad lasts longer than traditional cleaning pads
- Effective on most coated and uncoated hard floors including VCT, vinyl, rubber, marble, stone, terrazzo, granite, and concrete with only water or neutral cleaner

Specifications
Cleaning Pads | |
610 mm | |
Low Speed Scrubbers |
Brown | |
Cleaning and Polishing | |
2 in. 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction