Learn More
Sampling Systems™ Cleaning Coupons for Plastics
Test and validate your cleaning regime with this range of cleaning validation coupons for various plastics.
Brand:  Sampling Systems™ 8644PP-50
 View more versions of this product
Product Code. 15849551
  • 24.05 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Tests available for HDPE, nylon, PP, PTFE and PVC
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
Cleaning Coupon
50 x 50 mm
Testing and validating cleaning regime
Polypropylene