missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Type 1 Diagnostic Clear Glass Vials

Provides excellent clarity and dimensional accuracy from vial to vial due to tubular design. Fisherbrand™ Type 1 Diagnostic Clear Glass Vials are manufactured using Type 1 neutral glass.

Brand:  Fisherbrand™ 102080

151.05 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 14843582

  • 163.00 € / Pack of 99
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Freeze dry stoppers demonstrate very low moisture absorption
  • Specially designed bottom radius adds strength for freeze drying applications
  • Two cap configurations available - PP cap and freeze dry stopper urea EPE lined cap
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
Borosilicate Glass
Suitable for freeze drying applications, all diagnostic and pharmaceutical applications
Tubular
Autoclavable
99/Pk.
32 mm
25 mL
Screw
Screw
63 mm
Diagnostic Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers