Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates



Carry out colormetric assays with reliable and reproducible results using these clear, flat-bottom, immuno nonsterile 384-well plates, offered with a choice of surface.
272.70 € - 416.00 €
Specifications
Barcode | No Barcode |
---|---|
Binding Property | Hydrophilic |
Format | Standard |
Detection Method | Colorimetric |
Array | 16 x 24 |
Alert:
Each lot is functionally tested for IgG binding.
Description
Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates, offered with a choice of surface for optimal binding. Includes Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
384-Well Solid Plates:
- Clear polystyrene
- Choice of surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
- Thermo ScientificMaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
Ordering Information:
Please note that availability may vary by country.
Specifications
No Barcode | |
Standard | |
16 x 24 | |
Microplate | |
384 | |
Non-sterile | |
None |
Hydrophilic | |
Colorimetric | |
Clear | |
Polystyrene | |
100 μL | |
10 | |
Antibodies (Glycoproteins) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.