Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Brand: Thermo Scientific™ 456537
540.00 EUR valid until 2024-12-31
Use promo code "21894" to get your promotional price.
Alert:
Each lot is functionally tested for IgG binding.
Description
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of four surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
- Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
- Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
Specifications
No Barcode | |
Yes | |
Without Lid | |
8 x 12 | |
Hydrophilic | |
Microplate | |
400 μL | |
350 μL | |
MaxiSorp | |
180 | |
None | |
Antibodies (Glycoproteins) |
Hydrophilic | |
Standard | |
Colorimetric | |
180 Cases | |
Clear | |
Polystyrene | |
96 | |
Non-sterile | |
Round, Flat Bottom (F-well) | |
10 | |
180/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.