missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
BIOPHARMAGREENER_CHOICE

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.

Brand:  Thermo Scientific™ 439454

226.01 EUR valid until 2024-12-14
Use promo code "21632" to get your promotional price.


 View more versions of this product

Product Code. 10394751

  • 339.00 € / Pack of 60
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Alert:

Each lot is functionally tested for IgG binding.

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21632” to get your promotional price


Description

Description

Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No Barcode
Yes
Without Lid
8 x 12
Clear
Pinchbar
400 μL
350 μL
MaxiSorp
60
None
Antibodies (Glycoproteins)
Hydrophilic
Standard
Colorimetric
12 inner bags of 5 each
Microplate
Polystyrene
96
Non-sterile
Flat Bottom (F-well)
5
60/Cs.
F96
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers