missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
GREENER_CHOICE

Product Code. 10795026
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
MediSorp
MultiSorp
PolySorp
Universal Binding
Certified:
No
Yes
Volume (Metric) Working:
230 μL
280 μL
330 μL
350 μL
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 180
Pack of 50
Pack of 60
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Product Code. 10795026

Brand: Thermo Scientific™ 3455

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.

Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certified No
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 50 Cases
Binding Type Slightly Hydrophilic
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Volume (Metric) Working 280 μL
Sterility Non-sterile
Surface Treatment Immulon 2 HB
Well Shape Round, Flat Bottom (F-well)
No. per Case 50
No. per Pack 10
Plate Blocking None
Quantity 50/Pk.
Target Molecule Proteins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.