missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Clear PTFE Hoses

Kartell™ Clear PTFE Hoses have outstanding resistance to chemicals, which makes it suitable for the majority of fluid and ambient temperature conditions found in the food and pharmaceutical industry.

Brand:  Kartell™ 0390200

 View more versions of this product

Product Code. 10397981

  • 89.30 € / 5m
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Good resistance to acids, bases, and solvents
  • Can withstand temperatures up to 260°C
  • Autoclavable
Specifications

Specifications

4 x 6mm
4 mm
6 mm
260°C
4 x 6mm
260°C
Autoclavable
PTFE
5 m
1 Pack
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.