missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates
GREENER_CHOICE

Product Code. 10111005
missing translation for 'orderingAttributeHoverText'
Surface Treatment:
Immulon 1 B
Immulon 2 HB
MaxiSorp
PolySorp
missing translation for 'unitSize'
Pack of 50
Pack of 60
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Product Code. 10111005

missing translation for 'mfr': Thermo Scientific™ 3655

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Each lot is functionally tested for IgG binding.

Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.

Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces

  • PolySorp and Immulon 1B for adsorption of hydrophobic molecules
  • Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules
  • MaxiSorp is hydrophilic and ideal for antibody sandwich assays

Ordering Information:

Please note that availability may vary by country.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Slightly Hydrophilic
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 50 Cases
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 280 μL
No. of Wells 96
Volume (Metric) Working 180 μL
Sterility Non-sterile
Surface Treatment Immulon 2 HB
Well Shape Round
No. per Case 50
No. per Pack 10
Plate Blocking None
Target Molecule Proteins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.