Learn More
Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.
Brand: Thermo Scientific™ 449824
116.19 EUR valid until 2024-12-31
Use promo code "21894" to get your promotional price.
Alert:
Each lot is functionally tested for IgG binding.
Description
Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of a variety of surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp and Immulon 1B are for adsorption of hydrophobic molecules.
- Thermo Scientific Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules.
- Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays.
Ordering Information:
Please note that availability may vary by country.
Specifications
No Barcode | |
Standard | |
8 x 12 | |
Clear | |
Polystyrene | |
96 | |
Non-sterile | |
Round | |
5 | |
Antibodies (Glycoproteins) |
Hydrophilic | |
Colorimetric | |
60 Cases | |
Microplate | |
300 μL | |
250 μL | |
MaxiSorp | |
60 | |
None |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.