Learn More
Diversey ClearKlens ECO VH39 20L
Defoamed, organic acid detergent descaler for general CIP
Brand:  Diversey 101107534
Product Code. 17883105
  • 175.00 € / 20 L
Explore more special offers
Description
Description
  • Highly effective in removing organic and inorganic scale deposits, thus improving operational efficiency
  • Highly effective in removing many coating across the pharmaceutical industry
  • Its defoaming properties make it suitable for use under conditions of high pressure and turbulence, improving cleaning performance
  • Very low phosphors and nitrogen content, which means that it meets most of the customers environmental requirements across all borders
  • Suitable for automatic dosing and control by conductivity, ensuring consistent delivery of product
Specifications
Specifications
5.3 gal.
20 L