Learn More
Diversey™ ClearKlens IPA VH1 Sterile Disinfectant
Ready to use sterile surface disinfectant based on IPA 70%
208.00 € - 353.00 €
Specifications
Product Type | Surface Disinfectant |
---|---|
pH | 6 to 9 |
Sterility | Sterile |
Composition | 70% IPA/30% WFI USP |
Product Code | Brand | Volume (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Description | Price | Quantity & Availability | ||||
17633235
|
Diversey™
101104901 |
5 L | For ES, GB, IE, IT, PT only |
353.00 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17683235
|
Diversey™
101105062 |
900 mL | For ES, GB, IE, IT, PT, RU, SI only |
208.00 €
5.40 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17673235
|
Diversey™
101104905 |
1 L | For GB, AT, BE, CH, DE, FR, NL only |
327.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ClearKlens IPA is a ready to use disinfectant for the Life Sciences cleanroom industry. ClearKlens IPA is based on 70% isopropyl alcohol. ClearKlens IPA is available as 900 mL bag in bottle trigger spray, and as 1 L bottle or 5 L canister. The 900 mL bottle has been filtered to 0.2 μm and irradiated. The 1 L/5 L has been filtered to 0.2 μm and aseptically filled.
Key properties
- ClearKlens IPA is a solution of 70% isopropyl alcohol and 30% water for injection (USP quality).
- ClearKlens IPA is a ready to use product for the disinfection of work surfaces and process equipment in cleanrooms
- ClearKlens IPA 900 mL trigger spray is an antisuck back- bag in bottle delivery system to eliminate product contamination during use.
- BPR approved pack and formulation
- Double bagged for easy transfer into the cleanroom.
Benefits
- Ready to use
- 0.2 μm filtered
- Gamma irradiated/aseptically filled and double bagged
- Detex labelled for proof of irradiation
- Double bagged for easy transfer into the cleanroom
- Odorless and Non tainting
- Broad spectrum of activity
Specifications
Surface Disinfectant | |
6 to 9 | |
Sterile | |
70% IPA/30% WFI USP |
Safety and Handling
- ClearKlens IPA VH1
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Propan-2-ol
Each lot is provided with: Certificate of Analysis, Certificate of Sterility, Certificate of Irradiation
Your input is important to us. Please complete this form to provide feedback related to the content on this product.