Learn More
Diversey™ ClearKlens Oxifast Disinfectant
BIOPHARMA
Product Code. 17653095
Click to view available options
Volume (Metric):
1 L
Unit Size:
8 L
Product Code. 17653095
Brand: Diversey™ 100934612
ClearKlens Oxifast is designed for disinfecting hard surfaces and equipments in critical and non-critical areas.
  • ClearKlens Oxifast is used for the disinfection of walls, floors, surfaces, and process equipments in the Life Sciences industry.
  • ClearKlens Oxifast passes EN 13704 test with a shortened contact time of only 5 minutes, an important selection criterion for a Cleanroom disinfectant where wet
    contact times are limited.
Benefits
  • Ready to use sporicidal agent
  • Broad spectrum activity even at very short contact time: bactericidal, yeasticidal, fungicidal, virucidal, sporicidal
  • Low environmental impact
Specifications
Sterility Non-sterile
Composition Ready to use peracetic acid based solution
Quantity 8 Pk.
Volume (Metric) 1 L