missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit
GREENER_CHOICE

Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit

Brand:  Molecular Probes™ C10428

608.00 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 13532550

  • 800.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit provides a fast, sensitive, non-toxic, and non-radioactive method for the detection of nascent protein synthesis utilizing fluorescence microscopy, high-content imaging, or flow cytometry. Included in the kit are L-homopropargylglycine (HPG), an amino acid analog of methionine containing an alkyne moiety, and Alexa Fluor™ 488 azide. The HPG is fed to cultured cells and incorporated into proteins during active protein synthesis. Addition of the Alexa Fluor™ 488 azide leads to a chemoselective ligation or 'click' reaction between the green fluorescent azide and the alkyne, allowing the modified proteins to be detected by imaged-based analysis.

  • Non-radioactive alternative—an alternative to the traditional 35S-methionine
  • Visualize bulk protein dynamics—fluorescent tagging of proteins allows their localization to be determined, including aggregation
  • Specificity—selective, specific reaction between label and detection tags
  • Stability —product contains an irreversible, covalent bond
  • Multiplex-enabled—use in conjuction with Click™-iT AHA (azide amino acid and alkyne dye) to detect spatial and temporal differences
  • Applicability to biological samples—easy detection; high sensitivity and low background, regardless of complexity

    The Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit has been successfully tested in HeLa, A549, and U-2 OS cells with a variety of reagents that inhibit protein synthesis, including cycloheximide and anisomycin. The applicability of these probes to monitor protein degradation has also been shown using inhibitors of the proteasome (MG132 and Bortezomib) and blockers of autophagy (chloroquine) in HeLa cells.

    Additionally, due to differences in Click-iT™ chemistry between the Click-iT™ HPG Alexa Fluor™ 488 Protein Synthesis Assay Kit and Click™-iT AHA with Alexa Fluor™ 594 Alkyne, these reagents can be used in conjunction for spatial or temporal determination of differences in nascent protein synthesis.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Green
    Fluorescence
    Fluorescence Microscope, Fluorescent Imager
    Proteins (Nascent)
    Liquid
    Alexa Fluor™, Click-iT™
    • Store at 2°C to 6°C
    • DO NOT FREEZE
    • Protect material from long-term exposure to light, but may be exposed to light for short periods of time.
    Protein Synthesis Assay
    Protein Synthesis Assay Kit
    Visible
    Less hazardous
    1 kit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.