missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Click-iT™ Plus EdU Pacific Blue™ Flow Cytometry Assay Kit

Click-iT™ Plus EdU Pacific Blue™ Flow Cytometry Assay Kit

Brand:  Molecular Probes™ C10636

Product Code. 15342202

  • 708.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Click-iT™ Plus EdU Pacific Blue™ Flow Cytometry Assay Kit provides a simplified, more robust assay for analyzing DNA replication in proliferating cells compared to traditional BrdU methods. Newly synthesized DNA is analyzed using the 405 nm laser of the flow cytometer. The Click-iT™ Plus formulation is compatible with standard fluorophores, including R-PE and R-PE tandems, as well as fluorescent proteins.

  • Multiplexable—compatible with R-PE (and tandems) and fluorescent proteins
  • Accurate—superior results compared to BrdU assays
  • Fast—results in as little as 60 minutes

    View selection guide for all Click-iT™ EdU and Click-iT™ Plus EdU assays for flow cytometry.

    Multiplexable
    The Click-iT™ Plus formulation provides increased multiplexibility compared to the original Click-iT™ EdU Flow Cytometry assays. Click-iT™ Plus EdU assays can be used in conjunction with R-PE and R-PE tandems, as well as fluorescent proteins such as GFP and mCherry, without loss of the accuracy or speed of the original Click-iT™ EdU assay.

    An Advanced Method Giving You Results Superior to BrdU
    The most accurate method of proliferation analysis is direct measurement of DNA synthesis. Originally, this was performed through incorporation of radioactive nucleosides. This method was replaced by antibody-based detection of the nucleoside analog bromodeoxyuridine (BrdU). The Click-iT™ Plus EdU Flow Cytometry assay is a novel alternative to the BrdU assay. EdU (5-ethynyl-2´-deoxyuridine) is a thymidine analog that is incorporated into DNA during active DNA synthesis. Detection is based on click chemistry, which is a copper-catalyzed covalent reaction between an azide and an alkyne. In this application, the alkyne is found in the ethynyl moiety of EdU, while the azide is coupled to the Alexa Fluor™ dye. Standard flow cytometry methods are used for determining the percentage of S-phase cells in the population.

    Mild Conditions Allow Use with Cell Cycle Dyes and Antibodies
    The small size of the dye azide allows for efficient detection of the incorporated EdU using mild conditions, while standard aldehyde-based fixation and detergent permeabilization are sufficient for the Click-iT™ Plus detection reagent to gain access to the DNA. This is in contrast to BrdU assays that require DNA denaturation (using HCl, heat, or digestion with DNase) to expose the BrdU so that it can be detected with an anti-BrdU antibody. Sample processing for the BrdU assay can result in signal alteration of the cell cycle distribution, as well as destruction of antigen recognition sites when using the HCl method. In contrast, the easy-to-use Click-iT™ Plus EdU assay is compatible with cell cycle dyes. The Click-iT™ Plus EdU assay can also be multiplexed with antibodies against surface and intracellular markers, as well as conjugates labeled with standard fluorophores including R-PE, R-PE tandems, and fluorescent proteins (GFP and mCherry).

    Quick and Simple Protocol
    The Click-iT™ Plus EdU protocol is based on the aldehyde fixation and detergent permeabilization steps for immunohistochemical antibody labeling. However, EdU is compatible with other fixation/permeabilization agents including saponin and methanol. In just five steps you’ll be ready to analyze your cell proliferation data:
    1. Treat cells with EdU.
    2. Fix and permeabilize cells.
    3. Detect S-phase cells with Click-iT™ Plus detection cocktail for 30 min.
    4. Wash once.
    5. Analyze.
    Results can be seen in as little as 60 minutes in some circumstances, but we recommend 90 minutes for all applications.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Contains EdU (5-ethynyl-2' -deoxyuridine), Pacific Blue™ picolyl azide, anhydrous dimethylsulfoxide (DMSO), Click-iT™ fixative, Click-iT™ saponin-based permeabilization and wash buffer, Copper Protectant, and Click-iT™ EdU buffer additive.
  • Store at 2°C to 8°C
  • Dessicate and protect from light.
    Flow Cytometry
    Flow Cytometry Assay Kit
    404⁄450
    Click-iT™
    Room Temperature
    Fluorescence
    Flow Cytometer
    Pacific Blue™
    Tube(s)
    50 assays
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.